missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-methyl-L-serine, 97%

CAS: 51293-47-1 | C9H17NO5 | 219.237 g/mol

Supplier:  Thermo Scientific Chemicals H52829MD

 View more versions of this product

Catalog No. AAH52829MD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Boc-O-methyl-L-serine
C9H17NO5
boc-o-methyl-l-serine, boc-ser me-oh, s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid, boc-ser me-oh.dcha, n-boc-o-methyl-l-serine, s-2-tert-butoxycarbonylamino-3-methoxypropanoic acid, boc-ser me-oh dcha, boc-o-methyl-l-ser, serine, n-1,1-dimethylethoxy carbonyl-o-methyl, 2s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid
CC(C)(C)OC(=O)NC(COC)C(=O)O
250 mg
7016354
97%
51293-47-1
MFCD00153314
RFGMSGRWQUMJIR-LURJTMIESA-N
(2S)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
219.237
219.24

Chemical Identifiers

51293-47-1
219.237
RFGMSGRWQUMJIR-LURJTMIESA-N
7016354
CC(C)(C)OC(=O)NC(COC)C(=O)O
C9H17NO5
MFCD00153314
boc-o-methyl-l-serine, boc-ser me-oh, s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid, boc-ser me-oh.dcha, n-boc-o-methyl-l-serine, s-2-tert-butoxycarbonylamino-3-methoxypropanoic acid, boc-ser me-oh dcha, boc-o-methyl-l-ser, serine, n-1,1-dimethylethoxy carbonyl-o-methyl, 2s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid
(2S)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only