missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-benzyl-L-serine, 97%

Catalog No. AAH6270514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6270514 25 g
AAH6270506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6270514 Supplier Thermo Scientific Chemicals Supplier No. H6270514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23680-31-1 | C15H21NO5 | 295.335 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23680-31-1
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00066063
InChI Key DMBKPDOAQVGTST-LBPRGKRZSA-N
Synonym boc-ser bzl-oh, n-boc-o-benzyl-l-serine, boc-o-benzyl-l-serine, boc-ser bzl, s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-serine, n-t-butoxycarbonyl-o-benzyl-l-serine, o-benzyl-n-tert-butyloxycarbonylserine, n-tert-butoxycarbonyl-o-benzyl-l-serine, o-benzyl-n-tert-butoxycarbonyl-l-serine
PubChem CID 90234
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
SMILES CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 58°C to 60°C
Beilstein 3064461
Quantity 25 g
Formula Weight 295.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-O-benzyl-L-serine
EINECSNumber 245-820-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only