missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-benzyl-D-tyrosine, 95%

Catalog No. AAH6342714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6342714 25 g
AAH6342706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH6342714 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H6342714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63769-58-4 | C21H25NO5 | 371.43 g/mol

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 63769-58-4
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00038249
InChI Key ZAVSPTOJKOFMTA-LGWFVXIRNA-N
Synonym boc-d-tyr bzl-oh, boc-o-benzyl-d-tyrosine, n-boc-o-benzyl-d-tyrosine, boc-o-benzyl-d-tyr, r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, ambotzbaa1381, boc-dtyr bn-oh, pubchem14949, boc-d-tyr bzl oh
PubChem CID 6992575
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O

Specifications

Solubility Information Slightly soluble in water.
Quantity 25 g
Formula Weight 371.43
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-O-benzyl-D-tyrosine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only