missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-valine, 95%

CAS: 45170-31-8 | C11H21NO4 | 231.29 g/mol

$137.53 - $586.70

Chemical Identifiers

CAS 45170-31-8
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00038760
InChI Key XPUAXAVJMJDPDH-QMMMGPOBSA-N
Synonym boc-n-me-val-oh, boc-n-methyl-l-valine, n-boc-n-methyl-l-valine, boc-meval-oh, s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, ambotzbaa1272, n-boc-n-methylvaline, pubchem12254
PubChem CID 7010608
IUPAC Name (2S)-3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid
SMILES CC(C)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6277103
View Documents
Thermo Scientific Chemicals
H6277103
1 g
Each for $137.53
Only null left
 
AAH6277106
View Documents
Thermo Scientific Chemicals
H6277106
5 g
Each for $586.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

45170-31-8
231.29
XPUAXAVJMJDPDH-QMMMGPOBSA-N
7010608
CC(C)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
C11H21NO4
MFCD00038760
boc-n-me-val-oh, boc-n-methyl-l-valine, n-boc-n-methyl-l-valine, boc-meval-oh, s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, ambotzbaa1272, n-boc-n-methylvaline, pubchem12254
(2S)-3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid

Specifications

45170-31-8
MFCD00038760
boc-n-me-val-oh, boc-n-methyl-l-valine, n-boc-n-methyl-l-valine, boc-meval-oh, s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, ambotzbaa1272, n-boc-n-methylvaline, pubchem12254
CC(C)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
1 g
7010608
95%
N-Boc-N-methyl-L-valine
C11H21NO4
2646180
XPUAXAVJMJDPDH-QMMMGPOBSA-N
(2S)-3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid
231.29
231.29
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only