missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-alanine, 98%

Catalog No. AAH3131706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3131706 5 g
AAH3131703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3131706 Supplier Thermo Scientific Chemicals Supplier No. H3131706

CAS: 16948-16-6 | C9H17NO4 | 203.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16948-16-6
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037242
InChI Key VLHQXRIIQSTJCQ-LURJTMIESA-N
Synonym boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acid
PubChem CID 7009590
IUPAC Name (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES C[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 88°C to 92°C
Odor Odorless
Quantity 5 g
Optical Rotation −31° (c=1 in Ethanol)
Formula Weight 203.24
Percent Purity 98%
Chemical Name or Material N-Boc-N-methyl-L-alanine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only