missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-L-Valinol, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 399230050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC399230050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

79069-14-0
203.282
OOQRRYDVICNJGC-MRVPVSSYSA-N
7021464
CC(C)C(CO)NC(=O)OC(C)(C)C
C10H21NO3
MFCD00082635
n-boc-l-valinol, boc-l-valinol, boc-valinol, n-tert-butoxycarbonyl-l-valinol, s-n-tert-butoxycarbonyl valinol, r-n-tert-butoxycarbonyl valinol, tert-butyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, boc-valch2oh, n-t-boc-l-valinol
tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate

Specifications

N-BOC-L-Valinol
Glass bottle
100.0
5g
96%
MFCD00082635
OOQRRYDVICNJGC-MRVPVSSYSA-N
tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
7021464
96%
79069-14-0
95.0
0.9950g/mL
85°C
C10H21NO3
n-boc-l-valinol, boc-l-valinol, boc-valinol, n-tert-butoxycarbonyl-l-valinol, s-n-tert-butoxycarbonyl valinol, r-n-tert-butoxycarbonyl valinol, tert-butyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, boc-valch2oh, n-t-boc-l-valinol
CC(C)C(CO)NC(=O)OC(C)(C)C
203.282
203.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...