missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-tyrosine methyl ester, 99%, Thermo Scientific Chemicals

Catalog No. p-7026255
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2216414 25 g
AAB2216422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2216414 Supplier Thermo Scientific Chemicals Supplier No. B2216414

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4326-36-7
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00191181
InChI Key NQIFXJSLCUJHBB-STGVRZAANA-N
Synonym boc-tyr-ome, boc-l-tyrosine methyl ester, n-boc-l-tyrosine methyl ester, n-tert-butoxycarbonyl-l-tyrosine methyl ester, methyl n-boc-l-tyrosinate, methyl 2s-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, methyl 2s-2-tert-butoxy carbonyl amino-3-4-hydroxyphenyl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, s-2-tert-butoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid methyl ester
PubChem CID 7019130
IUPAC Name methyl (2S)-3-(4-hydroxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 105°C to 108°C
Quantity 25 g
Optical Rotation +50° (c=1 Chloroform)
Formula Weight 295.34
Percent Purity 99%
Chemical Name or Material N-Boc-L-tyrosine methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only