missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-BOC-L-Tryptophane, 97%

CAS: 13139-14-5 | C16H20N2O4 | 304.35 g/mol

$102.43 - $102.43

Chemical Identifiers

CAS 13139-14-5
Molecular Formula C16H20N2O4
Molecular Weight (g/mol) 304.35
MDL Number MFCD000655
InChI Key NFVNYBJCJGKVQK-ZDUSSCGKSA-N
Synonym boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
PubChem CID 83169
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC275690050
SDS View Documents
Thermo Scientific Chemicals
275690050
5 g Glass Bottle
Each for $102.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13139-14-5
304.35
NFVNYBJCJGKVQK-ZDUSSCGKSA-N
83169
C16H20N2O4
MFCD000655
boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

13139-14-5
100.0
Authentic
C16H20N2O4
MFCD000655
22, 6789
1% Max. (K.F.)
CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
5 g
83169
≥96.0%
Nα-BOC-L-Tryptophane
96.0
White
97%
Glass Bottle
-19° to -23° (20°C, 589nm) (c=1, CH3COOH)
boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
NFVNYBJCJGKVQK-ZDUSSCGKSA-N
(2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
304.35
304.34
Powder
Videos
SDS
Documents

Documents

RUO – Research Use Only