missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-L-tert-Leucine, 98%

CAS: 62965-35-9 | C11H21NO4 | 231.29 g/mol

$170.69 - $170.69

Chemical Identifiers

CAS 62965-35-9
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00065574
InChI Key LRFZIPCTFBPFLX-JLDDOWRYNA-N
Synonym n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine
PubChem CID 2734668
SMILES CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368010050
SDS View Documents
Thermo Scientific Chemicals
368010050
5 g Glass Bottle
Each for $170.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62965-35-9
231.29
LRFZIPCTFBPFLX-JLDDOWRYNA-N
2734668
C11H21NO4
MFCD00065574
n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C

Specifications

118.0°C to 121.0°C
97.5
White to Yellow
98%
Glass Bottle
-4.0° to -5.0° (20°C, 589nm) (c=1, acetic acid)
LRFZIPCTFBPFLX-JLDDOWRYNA-N
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
231.29
231.29
Crystalline Powder
62965-35-9
100.0
Authentic
C11H21NO4
MFCD00065574
n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C
5 g
2734668
98%
N-BOC-L-tert-Leucine
Videos
SDS
Documents

Documents

RUO – Research Use Only