missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-tert-leucine, 98%

Catalog No. AAH5113603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5113603 1 g
AAH5113606 5 g
AAH5113614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5113603 Supplier Thermo Scientific Chemicals Supplier No. H5113603

CAS: 62965-35-9 | C11H21NO4 | 231.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62965-35-9
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00065574
InChI Key LRFZIPCTFBPFLX-JLDDOWRYNA-N
Synonym n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine
PubChem CID 2734668
SMILES CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C

Specifications

Melting Point 118°C to 121°C
Quantity 1 g
IUPAC Name (2S)-3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Formula Weight 231.29
Percent Purity 98%
Chemical Name or Material N-Boc-L-tert-leucine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only