missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-serine methyl ester, 95%

Catalog No. AAH5988909
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5988909 10 g
AAH5988904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5988909 Supplier Thermo Scientific Chemicals Supplier No. H5988909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2766-43-0 | C9H17NO5 | 219.237 g/mol

N-tert-Butoxycarbonyl-L-serine Methyl Ester is an amino acid derivative used in the preparation of Tn antigen, L-glutamic acid p-nitroanilide analogs and new biomedical polymers having Serine and Threonine side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-tert-Butoxycarbonyl-L-serine Methyl Ester is an amino acid derivative used in the preparation of Tn antigen, L-glutamic acid p-nitroanilide analogs and new biomedical polymers having Serine and Threonine side groups.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 2766-43-0
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00191869
InChI Key SANNKFASHWONFD-LURJTMIESA-N
Synonym boc-ser-ome, boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, n-boc-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-l-ser-ome, methyl tert-butoxycarbonyl-l-serinate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-ser-ome; boc-l-serine methyl ester
PubChem CID 7019177
IUPAC Name methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

Density 1.082
Flash Point 113°C (235°F)
Refractive Index 1.4525
Linear Formula HOCH2CH[NHCO2C(CH3)3]CO2CH3
Quantity 10 g
Beilstein 3545389
Solubility Information Slightly soluble in water.
Optical Rotation −18° (c=5 in Methanol)
Formula Weight 219.23
Percent Purity 95%
Chemical Name or Material N-Boc-L-serine methyl ester
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only