missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-prolinal, 96%

Catalog No. AAH2649506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2649506 5 g
AAH2649503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2649506 Supplier Thermo Scientific Chemicals Supplier No. H2649506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69610-41-9 | C10H17NO3 | 199.25 g/mol

Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.

Solubility
Slightly soluble water. Soluble in chloroform.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.

Chemical Identifiers

CAS 69610-41-9
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00274186,MFCD00801206
InChI Key YDBPZCVWPFMBDH-QMMMGPOBSA-N
Synonym n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal
PubChem CID 2734674
IUPAC Name tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1C=O

Specifications

Density 1.06
Boiling Point 228°C
Flash Point >110°C (230°F)
Refractive Index 1.4589
Quantity 5 g
Sensitivity Air sensitive
Solubility Information Slightly soluble water. Soluble in chloroform.
Optical Rotation −95° (c=1 in Chloroform)
Formula Weight 199.25
Percent Purity 96%
Chemical Name or Material N-Boc-L-prolinal
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only