missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-pipecolinic acid, 98+%

Catalog No. AAL1936103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1936103 1 g
AAL1936106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1936103 Supplier Thermo Scientific Chemicals Supplier No. L1936103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26250-84-0 | C11H19NO4 | 229.276 g/mol

N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 26250-84-0
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00151904
InChI Key JQAOHGMPAAWWQO-QMMMGPOBSA-N
Synonym boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid
PubChem CID 688617
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

Melting Point 121°C to 125°C
Quantity 1 g
Beilstein 3652038
Solubility Information Insoluble in water.
Optical Rotation −46° (c=1 in Methanol)
Formula Weight 229.28
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-pipecolinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only