missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalanyl-L-phenylalanine, 95%

Catalog No. AAH6378806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6378806 5 g
AAH6378803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6378806 Supplier Thermo Scientific Chemicals Supplier No. H6378806

CAS: 13122-90-2 | C23H28N2O5 | 412.486 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13122-90-2
Molecular Formula C23H28N2O5
Molecular Weight (g/mol) 412.486
MDL Number MFCD00190824
InChI Key NNOBHAOOLCEJBL-OALUTQOASA-N
Synonym boc-phe-phe-oh, boc-l-diphenylalanine, s-2-s-2-tert-butoxycarbonyl amino-3-phenylpropanamido-3-phenylpropanoic acid, n-tert-butoxycarbonyl-l-phenylalanyl-l-phenylalanine, 2s-2-2s-2-tert-butoxycarbonyl amino-3-phenylpropanamido-3-phenylpropanoic acid, n-boc-l-phenylalanyl-l-phenylalanine, n-n-tert-butoxycarbonyl-l-phenylalanyl-l-phenylalanine
PubChem CID 7019052
IUPAC Name (2S)-2-[[(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoyl]amino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC2=CC=CC=C2)C(=O)O

Specifications

Quantity 5 g
Formula Weight 412.48
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-L-phenylalanyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only