missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalanine methyl ester, 95%

Catalog No. p-7054691
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6307806 5 g
AAH6307814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6307806 Supplier Thermo Scientific Chemicals Supplier No. H6307806

CAS: 51987-73-6 | C15H21NO4 | 279.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51987-73-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00076977
InChI Key SDSWSVBXRBXPRL-UHFFFAOYNA-N
Synonym boc-phe-ome, boc-l-phenylalanine methyl ester, l-boc-phenyl-alanine methyl ester, n-boc-l-phenylalanine methyl ester, n-tert-butoxycarbonyl-l-phenylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, s-2-tert-butoxycarbonylamino-3-phenyl-propionic acid methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, t-boc-l-phe-ome, pubchem13507
PubChem CID 9882138
IUPAC Name methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
SMILES COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 36°C to 40°C
Color White
Beilstein 3061910
Quantity 5 g
Flash Point 113°C (235°F)
Formula Weight 379.33
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-L-phenylalanine methyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only