missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-leucine hydrate, 99%, Thermo Scientific Chemicals

$44.30 - $125.70

Chemical Identifiers

CAS 13139-15-6
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00066067
InChI Key MDXGYYOJGPFFJL-SVGMAFHSNA-N
Synonym boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine
PubChem CID 83170
ChEBI CHEBI:28702
IUPAC Name (2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1064706
View Documents
Thermo Scientific Chemicals
A1064706
5 g
Each for $44.30
Only null left
 
AAA1064714
View Documents
Thermo Scientific Chemicals
A1064714
25 g
Each for $125.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13139-15-6
231.29
MDXGYYOJGPFFJL-SVGMAFHSNA-N
83170
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
C11H21NO4
MFCD00066067
boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine
CHEBI:28702
CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

13139-15-6
C11H21NO4
5 g
boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine
−25° (c=2 in acetic acid)
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
83170
231.29 (Anhydrous)
N-Boc-L-leucine hydrate
84°C to 87°C
MFCD00066067
1711814
MDXGYYOJGPFFJL-SVGMAFHSNA-N
CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O
231.29
CHEBI:28702
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 236-073-2

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only