missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-histidine methyl ester, 95%

CAS: 2488-14-4 | C12H19N3O4 | 269.301 g/mol

Supplier:  Thermo Scientific Chemicals H6254506

 View more versions of this product

Catalog No. AAH6254506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Boc-L-histidine methyl ester
C12H19N3O4
boc-his-ome, nalpha-boc-l-histidine methyl ester, l-histidine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, n-boc-l-histidine methyl ester, na-boc-l-histidine methyl ester, n-tert-butoxycarbonyl-l-histidine methyl ester, l-histidine,n-1,1-dimethylethoxy carbonyl-, methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-5-yl propanoate
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)OC
5 g
7017200
95%
2488-14-4
MFCD00167635
RWEVEEORUORBAX-VIFPVBQESA-N
methyl (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
269.301
269.3
Solid

Chemical Identifiers

2488-14-4
269.301
RWEVEEORUORBAX-VIFPVBQESA-N
7017200
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)OC
C12H19N3O4
MFCD00167635
boc-his-ome, nalpha-boc-l-histidine methyl ester, l-histidine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, n-boc-l-histidine methyl ester, na-boc-l-histidine methyl ester, n-tert-butoxycarbonyl-l-histidine methyl ester, l-histidine,n-1,1-dimethylethoxy carbonyl-, methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-5-yl propanoate
methyl (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only