missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-glutamic acid 1,5-dimethyl ester, 97%
CAS: 59279-60-6 | C12H21NO6 | 275.301 g/mol
$127.10 - $522.29
Chemical Identifiers
| CAS | 59279-60-6 |
|---|---|
| Molecular Formula | C12H21NO6 |
| Molecular Weight (g/mol) | 275.301 |
| MDL Number | MFCD03094774 |
| InChI Key | QNSPKWUAZQIIGZ-QMMMGPOBSA-N |
| Synonym | boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester |
| PubChem CID | 2734644 |
| IUPAC Name | dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate |
| SMILES | CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6205606
|
Thermo Scientific Chemicals
H6205606 |
5 g |
N/A
|
|
|||||
|
AAH6205614
|
Thermo Scientific Chemicals
H6205614 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 59279-60-6 | |
| 275.301 | |
| QNSPKWUAZQIIGZ-QMMMGPOBSA-N | |
| 2734644 | |
| CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC |
| C12H21NO6 | |
| MFCD03094774 | |
| boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester | |
| dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate |
Specifications
| 59279-60-6 | |
| C12H21NO6 | |
| boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester | |
| CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC | |
| 5 g | |
| 2734644 | |
| 97% | |
| N-Boc-L-glutamic acid 1,5-dimethyl ester |
| Colorless to Yellow | |
| MFCD03094774 | |
| QNSPKWUAZQIIGZ-QMMMGPOBSA-N | |
| dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate | |
| 275.301 | |
| 275.3 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only