missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 1,5-dimethyl ester, 97%

Catalog No. AAH6205606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6205606 5 g
AAH6205614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6205606 Supplier Thermo Scientific Chemicals Supplier No. H6205606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59279-60-6 | C12H21NO6 | 275.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59279-60-6
Molecular Formula C12H21NO6
Molecular Weight (g/mol) 275.301
MDL Number MFCD03094774
InChI Key QNSPKWUAZQIIGZ-QMMMGPOBSA-N
Synonym boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester
PubChem CID 2734644
IUPAC Name dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC

Specifications

Color Colorless to Yellow
Quantity 5 g
Formula Weight 275.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Boc-L-glutamic acid 1,5-dimethyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only