missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-beta-proline, 95%, Thermo Scientific Chemicals
$76.79 - $210.07
Chemical Identifiers
| CAS | 140148-70-5 |
|---|---|
| Molecular Formula | C10H17NO4 |
| Molecular Weight (g/mol) | 215.249 |
| MDL Number | MFCD03094728 |
| InChI Key | HRMRQBJUFWFQLX-ZETCQYMHSA-N |
| Synonym | s-1-boc-pyrrolidine-3-carboxylic acid, s-1-n-boc-beta-proline, s-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-l-beta-proline, 3s-1-tert-butoxycarbonyl-3-pyrrolidinecarboxylic acid, s-n-boc-pyrrolidine-3-carboxylic acid, 3s-boc-beta-pro-oh, 3s-1-boc-pyrrolidine-3-carboxylic acid, boc-3s-1-pyrrolidine-3-carboxylic acid |
| PubChem CID | 1512492 |
| IUPAC Name | (3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)N1CCC(C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH57888MD
|
Thermo Scientific Chemicals
H57888MD |
250 mg |
Each for $76.79
|
|
|||||
|
AAH5788803
|
Thermo Scientific Chemicals
H5788803 |
1 g |
Each for $210.07
|
|
|||||
Description
N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (slightly).
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 140148-70-5 | |
| 215.249 | |
| HRMRQBJUFWFQLX-ZETCQYMHSA-N | |
| 1512492 | |
| CC(C)(C)OC(=O)N1CCC(C1)C(=O)O |
| C10H17NO4 | |
| MFCD03094728 | |
| s-1-boc-pyrrolidine-3-carboxylic acid, s-1-n-boc-beta-proline, s-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-l-beta-proline, 3s-1-tert-butoxycarbonyl-3-pyrrolidinecarboxylic acid, s-n-boc-pyrrolidine-3-carboxylic acid, 3s-boc-beta-pro-oh, 3s-1-boc-pyrrolidine-3-carboxylic acid, boc-3s-1-pyrrolidine-3-carboxylic acid | |
| (3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid |
Specifications
| 140148-70-5 | |
| MFCD03094728 | |
| s-1-boc-pyrrolidine-3-carboxylic acid, s-1-n-boc-beta-proline, s-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-l-beta-proline, 3s-1-tert-butoxycarbonyl-3-pyrrolidinecarboxylic acid, s-n-boc-pyrrolidine-3-carboxylic acid, 3s-boc-beta-pro-oh, 3s-1-boc-pyrrolidine-3-carboxylic acid, boc-3s-1-pyrrolidine-3-carboxylic acid | |
| HRMRQBJUFWFQLX-ZETCQYMHSA-N | |
| CC(C)(C)OC(=O)N1CCC(C1)C(=O)O | |
| 215.249 | |
| 215.25 | |
| N-Boc-L-beta-proline |
| C10H17NO4 | |
| 250 mg | |
| Soluble in water (slightly). | |
| +15° (c=0.8 in Chloroform) | |
| (3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid | |
| 1512492 | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only