missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%

Catalog No. p-7034505
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52191MD 250 mg
AAH5219103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52191MD Supplier Thermo Scientific Chemicals Supplier No. H52191MD

CAS: 254101-10-5 | C17H23NO6 | 337.372 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 254101-10-5
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.372
MDL Number MFCD01862861
InChI Key FAFJSSKTLCNWRJ-CYBMUJFWSA-N
Synonym boc-beta-hoasp obzl-oh, boc-l-beta-glutamic acid 5-benzyl ester, boc-beta-glu obzl-oh, boc-, a-hoasp obzl-oh, r-5-benzyloxy-3-tert-butoxycarbonyl amino-5-oxopentanoic acid, 3r-5-benzyloxy-3-tert-butoxycarbonyl amino-5-oxopentanoic acid, fmoc-asparaginol, boc-beta-homoaspartic acid obzl, ambotzbaa6150, boc-hoasp obzl-oh
PubChem CID 2761513
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)CC(=O)OCC1=CC=CC=C1

Specifications

Beilstein 8430789
Quantity 250 mg
Formula Weight 337.37
Percent Purity 95%
Chemical Name or Material N-Boc-L-beta-glutamic acid 5-benzyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only