missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid 4-benzyl ester, 98%, Thermo Scientific Chemicals

Catalog No. p-7025994
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2175806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2175806 Supplier Thermo Scientific Chemicals Supplier No. B2175806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7536-58-5 | Molecular Formula: C16H21NO6 | Molecular Weight: 323.345 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7536-58-5
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00065564
InChI Key SOHLZANWVLCPHK-LBPRGKRZSA-N
Synonym boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
PubChem CID 1581888
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 110°C to 112°C
Quantity 5 g
Formula Weight 323.35
Percent Purity 98%
Chemical Name or Material N-Boc-L-aspartic acid 4-benzyl ester
EINECSNumber 231-406-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only