missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alaninol, 99%

Catalog No. AAB2239914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2239914 25 g
AAB2239906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2239914 Supplier Thermo Scientific Chemicals Supplier No. B2239914

CAS: 79069-13-9 | C8H17NO3 | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79069-13-9
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00043121
InChI Key PDAFIZPRSXHMCO-LURJTMIESA-N
Synonym boc-l-alaninol, n-boc-l-alaninol, boc-alaninol, s-2-boc-amino-1-propanol, n-tert-butoxycarbonyl-l-alaninol, s-tert-butyl 1-hydroxypropan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1-propanol, tert-butyl n-2s-1-hydroxypropan-2-yl carbamate, s---2-tert-butoxycarbonylamino-1-propanol
PubChem CID 7023103
IUPAC Name tert-butyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@@H](CO)NC(=O)OC(C)(C)C

Specifications

Melting Point 60°C to 62°C
Beilstein 3648840
Quantity 25 g
Optical Rotation −8.6° (c=1.2 in Methanol)
Formula Weight 175.23
Percent Purity 99%
Chemical Name or Material N-Boc-L-alaninol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only