missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alanine methyl ester, 95%

Catalog No. p-7054233
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6227106 5 g
AAH6227114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6227106 Supplier Thermo Scientific Chemicals Supplier No. H6227106

CAS: 28875-17-4 | C9H17NO4 | 203.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28875-17-4
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00038513
InChI Key GJDICGOCZGRDFM-LURJTMIESA-N
Synonym boc-ala-ome, boc-l-alanine methyl ester, s-n-tert-butoxycarbonylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino propanoate, n-boc-l-alanine methyl ester, boc-alanine methyl ester, s-methyl 2-tert-butoxycarbonylamino propanoate, methyl n-tert-butoxycarbonyl-l-alaninate, methyl 2s-2-tert-butoxycarbonyl amino propanoate, methyl 2s-2-tert-butoxy carbonyl amino propanoate
PubChem CID 10856577
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}propanoate
SMILES COC(=O)[C@H](C)NC(=O)OC(C)(C)C

Specifications

Melting Point 32°C to 35°C
Color White
Beilstein 1872545
Density 1.030 g/mL
Quantity 5 g
Flash Point 113°C (235°F)
Formula Weight 203.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-L-alanine methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only