Learn More
N-Boc-iminodiacetic acid, 98%
CAS: 56074-20-5 | C9H15NO6 | 233.22 g/mol
$129.59 - $501.34
Chemical Identifiers
| CAS | 56074-20-5 |
|---|---|
| Molecular Formula | C9H15NO6 |
| Molecular Weight (g/mol) | 233.22 |
| MDL Number | MFCD00800351 |
| InChI Key | QPGIJQUTGABQLQ-UHFFFAOYSA-N |
| Synonym | n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid |
| PubChem CID | 2755971 |
| IUPAC Name | 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid |
| SMILES | CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6686806
|
Thermo Scientific Chemicals
H6686806 |
5 g |
Each for $129.59
|
|
|||||
|
AAH6686814
|
Thermo Scientific Chemicals
H6686814 |
25 g |
Each for $501.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56074-20-5 | |
| 233.22 | |
| QPGIJQUTGABQLQ-UHFFFAOYSA-N | |
| 2755971 | |
| CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O |
| C9H15NO6 | |
| MFCD00800351 | |
| n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid | |
| 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid |
Specifications
| 56074-20-5 | |
| C9H15NO6 | |
| MFCD00800351 | |
| n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid | |
| CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O | |
| 5 g | |
| 2755971 | |
| 98% | |
| N-Boc-iminodiacetic acid |
| White | |
| (CH2CO2H)2NCOOC(CH3)3 | |
| 4435233 | |
| QPGIJQUTGABQLQ-UHFFFAOYSA-N | |
| 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid | |
| 233.22 | |
| 233.22 | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only