missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-iminodiacetic acid, 98%

Catalog No. AAH6686806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6686806 5 g
AAH6686814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6686806 Supplier Thermo Scientific Chemicals Supplier No. H6686806

CAS: 56074-20-5 | C9H15NO6 | 233.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56074-20-5
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00800351
InChI Key QPGIJQUTGABQLQ-UHFFFAOYSA-N
Synonym n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid
PubChem CID 2755971
IUPAC Name 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O

Specifications

Color White
Linear Formula (CH2CO2H)2NCOOC(CH3)3
Beilstein 4435233
Quantity 5 g
Formula Weight 233.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-iminodiacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only