missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-hydroxylamine, 98+%
Description
N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, reducing agents and alkali metals. Store in a cool place.
N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, reducing agents and alkali metals. Store in a cool place.
Specifications
Specifications
| Melting Point | 55°C to 58°C |
| Quantity | 1 g |
| Beilstein | 1756546 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 133.15 |
| Percent Purity | ≥98% |
| Chemical Name or Material | N-Boc-hydroxylamine |
Safety and Handling
| EINECSNumber | 252-836-2 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only