missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-hydroxylamine, 98+%

Catalog No. AAA1034906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1034906 5 g
AAA1034903 1 g
AAA1034914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1034906 Supplier Thermo Scientific Chemicals Supplier No. A1034906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 36016-38-3 | C5H11NO3 | 133.15 g/mol

N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, reducing agents and alkali metals. Store in a cool place.

Chemical Identifiers

CAS 36016-38-3
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002107
InChI Key DRDVJQOGFWAVLH-UHFFFAOYSA-N
Synonym n-boc-hydroxylamine, n-tert-butoxycarbonylhydroxylamine, tert-butyl hydroxycarbamate, boc-amino alcohol, carbamic acid, hydroxy-, 1,1-dimethylethyl ester, n-hydroxycarbamic acid tert-butyl ester, tert-butyl-n-hydroxycarbamate, n-tert-boc-hydroxylamine, t-butyl n-hydroxycarbamate, n-tert-butyloxycarbonylhydroxylamine
PubChem CID 97534
IUPAC Name tert-butyl N-hydroxycarbamate
SMILES CC(C)(C)OC(=O)NO

Specifications

Melting Point 55°C to 58°C
Quantity 5 g
Beilstein 1756546
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material N-Boc-hydroxylamine
EINECSNumber 252-836-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only