missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-guanidine, 95%

Catalog No. p-7054792
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6320606 5 g
AAH6320614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6320606 Supplier Thermo Scientific Chemicals Supplier No. H6320606

CAS: 219511-71-4 | C6H13N3O2 | 159.189 g/mol

N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.

Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 219511-71-4
Molecular Formula C6H13N3O2
Molecular Weight (g/mol) 159.189
MDL Number MFCD20547465
InChI Key UMOZLQVSOVNSCA-UHFFFAOYSA-N
Synonym boc-guanidine, n-boc-guanidine, 1-tert-butoxycarbonyl guanidine, tert-butyl n-carbamimidoylcarbamate, boc guanidine, 1-boc-guanidine, tert-butyl n-diaminomethylidene carbamate, tert-butoxycarbonylguanidine, acmc-209x9z
PubChem CID 11378649
IUPAC Name tert-butyl N-(diaminomethylidene)carbamate
SMILES CC(C)(C)OC(=O)N=C(N)N

Specifications

Melting Point 170°C to 175°C
Color White
Linear Formula (CH3)3OCONHC(=NH)NH2
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in most alcohols,esters (e.g.,ethyl acetate),ketones (e.g.,acetone),acetonitrile,and dichloromethane. Not soluble in diethyl ether and hexane.
Formula Weight 159.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-guanidine
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only