missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-glycine methyl ester, 97%
CAS: 31954-27-5 | C8H15NO4 | 189.211 g/mol
$82.11 - $231.26
Chemical Identifiers
| CAS | 31954-27-5 |
|---|---|
| Molecular Formula | C8H15NO4 |
| Molecular Weight (g/mol) | 189.211 |
| MDL Number | MFCD00038267 |
| InChI Key | PHUZOEOLWIHIKH-UHFFFAOYSA-N |
| Synonym | boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate |
| PubChem CID | 637609 |
| IUPAC Name | methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate |
| SMILES | CC(C)(C)OC(=O)NCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH59234AA
|
Thermo Scientific Chemicals
H59234AA |
5 mL |
Each for $82.11
|
|
|||||
|
AAH59234AC
|
Thermo Scientific Chemicals
H59234AC |
25 mL |
Each for $231.26
|
|
|||||
Description
It is used as a pharmaceutical, chemical, paint and dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical, chemical, paint and dye intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
| 31954-27-5 | |
| 189.211 | |
| PHUZOEOLWIHIKH-UHFFFAOYSA-N | |
| 637609 | |
| CC(C)(C)OC(=O)NCC(=O)OC |
| C8H15NO4 | |
| MFCD00038267 | |
| boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate | |
| methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate |
Specifications
| 31954-27-5 | |
| 190°C | |
| C8H15NO4 | |
| 1.437 | |
| 5 mL | |
| PHUZOEOLWIHIKH-UHFFFAOYSA-N | |
| methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate | |
| 637609 | |
| 97% |
| 1.079 | |
| 109°C (228°F) | |
| (CH3)3COCONHCH2CO2CH3 | |
| MFCD00038267 | |
| boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate | |
| CC(C)(C)OC(=O)NCC(=O)OC | |
| 189.211 | |
| 189.21 | |
| N-Boc-glycine methyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only