missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine methyl ester, 97%

Catalog No. AAH59234AA
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59234AA 5 mL
AAH59234AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59234AA Supplier Thermo Scientific Chemicals Supplier No. H59234AA

CAS: 31954-27-5 | C8H15NO4 | 189.211 g/mol

It is used as a pharmaceutical, chemical, paint and dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical, chemical, paint and dye intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 31954-27-5
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00038267
InChI Key PHUZOEOLWIHIKH-UHFFFAOYSA-N
Synonym boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate
PubChem CID 637609
IUPAC Name methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CC(C)(C)OC(=O)NCC(=O)OC

Specifications

Density 1.079
Boiling Point 190°C
Flash Point 109°C (228°F)
Refractive Index 1.437
Linear Formula (CH3)3COCONHCH2CO2CH3
Quantity 5 mL
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material N-Boc-glycine methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only