missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine, 98+%

CAS: 4530-20-5 | C7H13NO4 | 175.18 g/mol

Supplier:  Thermo Scientific Chemicals A1157914

 View more versions of this product

Catalog No. AAA1157914


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

87°C to 90°C
4530-20-5
MFCD00002690
boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
CC(C)(C)OC(=O)NCC(O)=O
25 g
78288
≥98%
N-Boc-glycine
C7H13NO4
1101514
VRPJIFMKZZEXLR-UHFFFAOYSA-N
2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
175.18
175.18

Chemical Identifiers

4530-20-5
175.18
VRPJIFMKZZEXLR-UHFFFAOYSA-N
78288
CC(C)(C)OC(=O)NCC(O)=O
C7H13NO4
MFCD00002690
boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

EINECSNumber : 224-864-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only