missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine, 98+%

Catalog No. AAA1157906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1157906 5 g
AAA1157914 25 g
AAA1157922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1157906 Supplier Thermo Scientific Chemicals Supplier No. A1157906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4530-20-5 | C7H13NO4 | 175.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4530-20-5
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD00002690
InChI Key VRPJIFMKZZEXLR-UHFFFAOYSA-N
Synonym boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
PubChem CID 78288
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)NCC(O)=O

Specifications

Melting Point 87°C to 90°C
Beilstein 1101514
Quantity 5 g
Formula Weight 175.18
Percent Purity ≥98%
Chemical Name or Material N-Boc-glycine
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 224-864-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only