missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-gamma-aminobutyric acid, 98+%
CAS: 57294-38-9 | C9H17NO4 | 203.24 g/mol
$63.38 - $261.26
Chemical Identifiers
| CAS | 57294-38-9 |
|---|---|
| Molecular Formula | C9H17NO4 |
| Molecular Weight (g/mol) | 203.24 |
| MDL Number | MFCD00037313 |
| InChI Key | HIDJWBGOQFTDLU-UHFFFAOYSA-N |
| Synonym | boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid |
| PubChem CID | 294894 |
| IUPAC Name | 4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2240703
|
Thermo Scientific Chemicals
B2240703 |
1 g |
Each for $63.38
|
|
|||||
|
AAB2240706
|
Thermo Scientific Chemicals
B2240706 |
5 g |
Each for $261.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 57294-38-9 | |
| 203.24 | |
| HIDJWBGOQFTDLU-UHFFFAOYSA-N | |
| 294894 | |
| CC(C)(C)OC(=O)NCCCC(O)=O |
| C9H17NO4 | |
| MFCD00037313 | |
| boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid | |
| 4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Specifications
| 57°C to 60°C | |
| C9H17NO4 | |
| boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid | |
| CC(C)(C)OC(=O)NCCCC(O)=O | |
| 1 g | |
| 294894 | |
| ≥98% |
| 57294-38-9 | |
| MFCD00037313 | |
| HIDJWBGOQFTDLU-UHFFFAOYSA-N | |
| 4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
| 203.24 | |
| 203.24 | |
| N-Boc-gamma-aminobutyric acid |
Safety and Handling
EINECSNumber : 628-652-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only