missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-ethylenediamine hydrochloride, 98%

Catalog No. p-7047590
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1948503 1 g
AAL1948506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1948503 Supplier Thermo Scientific Chemicals Supplier No. L1948503

CAS: 79513-35-2 | C7H17ClN2O2 | 196.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79513-35-2
Molecular Formula C7H17ClN2O2
Molecular Weight (g/mol) 196.68
MDL Number MFCD01076129
InChI Key XUHJJLCKXZTUJN-UHFFFAOYSA-N
Synonym n-boc-ethylenediamine hydrochloride, tert-butyl 2-aminoethyl carbamate hydrochloride, tert-butyl n-2-aminoethyl carbamate hydrochloride, n-boc-ethane-1,2-diaminehcl, n-boc-ethane-1,2-diamine hcl, n-tert-butoxycarbonylethylenediamine hydrochloride, n-tert-butoxycarbonyl-ethylenediamine hydrochloride, n-boc-ethylenediamine hcl, tert-butyl n-2-aminoethyl carbamate-hcl, n-boc-ethylenediaminehydrochloride
PubChem CID 187200
IUPAC Name tert-butyl N-(2-aminoethyl)carbamate;hydrochloride
SMILES [H+].[Cl-].CC(C)(C)OC(=O)NCCN

Specifications

Melting Point 148°C to 151°C
Beilstein 3627511
Quantity 1 g
Formula Weight 196.68
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material N-Boc-ethylenediamine hydrochloride
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only