missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-ethylenediamine hydrochloride, 98%

CAS: 79513-35-2 | C7H17ClN2O2 | 196.68 g/mol

Supplier:  Thermo Scientific Chemicals L1948506

 View more versions of this product

Catalog No. AAL1948506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

148°C to 151°C
79513-35-2
MFCD01076129
n-boc-ethylenediamine hydrochloride, tert-butyl 2-aminoethyl carbamate hydrochloride, tert-butyl n-2-aminoethyl carbamate hydrochloride, n-boc-ethane-1,2-diaminehcl, n-boc-ethane-1,2-diamine hcl, n-tert-butoxycarbonylethylenediamine hydrochloride, n-tert-butoxycarbonyl-ethylenediamine hydrochloride, n-boc-ethylenediamine hcl, tert-butyl n-2-aminoethyl carbamate-hcl, n-boc-ethylenediaminehydrochloride
[H+].[Cl-].CC(C)(C)OC(=O)NCCN
5 g
187200
Hygroscopic
N-Boc-ethylenediamine hydrochloride
C7H17ClN2O2
3627511
XUHJJLCKXZTUJN-UHFFFAOYSA-N
tert-butyl N-(2-aminoethyl)carbamate;hydrochloride
196.68
196.68
98%

Chemical Identifiers

79513-35-2
196.68
XUHJJLCKXZTUJN-UHFFFAOYSA-N
187200
[H+].[Cl-].CC(C)(C)OC(=O)NCCN
C7H17ClN2O2
MFCD01076129
n-boc-ethylenediamine hydrochloride, tert-butyl 2-aminoethyl carbamate hydrochloride, tert-butyl n-2-aminoethyl carbamate hydrochloride, n-boc-ethane-1,2-diaminehcl, n-boc-ethane-1,2-diamine hcl, n-tert-butoxycarbonylethylenediamine hydrochloride, n-tert-butoxycarbonyl-ethylenediamine hydrochloride, n-boc-ethylenediamine hcl, tert-butyl n-2-aminoethyl carbamate-hcl, n-boc-ethylenediaminehydrochloride
tert-butyl N-(2-aminoethyl)carbamate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only