missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-proline, 98%

Catalog No. p-7031314
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3234703 1 g
AAH3234706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3234703 Supplier Thermo Scientific Chemicals Supplier No. H3234703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59433-50-0 | C10H17NO4 | 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59433-50-0
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00520915
InChI Key ZQEBQGAAWMOMAI-UHFFFAOYSA-N
Synonym boc-dl-pro-oh, 1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl proline, n-boc-dl-proline, boc-dl-proline, 1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 1,2-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl ester, 1-2-methylpropan-2-yl oxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid
PubChem CID 268472
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O

Specifications

Melting Point 101°C to 104°C
Quantity 1 g
Formula Weight 215.25
Percent Purity 98%
Chemical Name or Material N-Boc-DL-proline
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only