missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-phenylglycine, 98%

Catalog No. p-7041500
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6235406 5 g
AAH6235414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6235406 Supplier Thermo Scientific Chemicals Supplier No. H6235406

CAS: 3601-66-9 | C13H17NO4 | 251.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3601-66-9
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.282
MDL Number MFCD00971063
InChI Key HOBFSNNENNQQIU-UHFFFAOYSA-N
Synonym boc-dl-phg-oh, tert-butoxycarbonylamino-phenyl-acetic acid, 2-tert-butoxycarbonyl amino-2-phenylacetic acid, tert-butoxycarbonyl amino phenyl acetic acid, n-boc-dl-phenylglycine, boc-phenylglycine, 2-tert-butoxy carbonyl amino-2-phenylacetic acid, rarechem ak ml 0503, 2-tert-butoxycarbonylamino-2-phenylacetic acid, boc-d-alpha-phenylglycine
PubChem CID 571805
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
SMILES CC(C)(C)OC(=O)NC(C1=CC=CC=C1)C(=O)O

Specifications

Quantity 5 g
Formula Weight 251.28
Percent Purity 98%
Chemical Name or Material N-Boc-DL-phenylglycine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only