missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-phenylalanine, 95%

Catalog No. AAH6316406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6316406 5 g
AAH6316414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6316406 Supplier Thermo Scientific Chemicals Supplier No. H6316406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4530-18-1 | C14H19NO4 | 265.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4530-18-1
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00558969
InChI Key ZYJPUMXJBDHSIF-UHFFFAOYSA-N
Synonym boc-dl-phenylalanine, boc-dl-phe-oh, n-boc-dl-phenylalanine, n-tert-butoxycarbonyl phenylalanine, 2-tert-butoxy carbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonylamino-3-phenylpropanoic acid, 2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoic acid, n-t-boc-phenylalanine
PubChem CID 269698
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)O

Specifications

Quantity 5 g
Formula Weight 265.3
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-DL-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only