missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-methionine, 98%

CAS: 93000-03-4 | C10H19NO4S | 249.33 g/mol

$111.58 - $432.49

Chemical Identifiers

CAS 93000-03-4
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.33
MDL Number MFCD00190811
InChI Key IMUSLIHRIYOHEV-UHFFFAOYNA-N
Synonym boc-dl-met-oh, boc-dl-methionine, methionine,n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl methionine, 2-tert-butoxy carbonyl amino-4-methylsulfanyl butanoic acid, 2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoic acid, n-alpha-t-boc-l-methionine, n-tert-butoxycarbonyl-d-methionine, tert-butoxycarbonyl-l-methionine, pubchem11999
PubChem CID 102827
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
SMILES CSCCC(NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6637203
View Documents
Thermo Scientific Chemicals
H6637203
1 g
Each for $111.58
Only null left
 
AAH6637206
View Documents
Thermo Scientific Chemicals
H6637206
5 g
Each for $432.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

93000-03-4
249.33
IMUSLIHRIYOHEV-UHFFFAOYNA-N
102827
CSCCC(NC(=O)OC(C)(C)C)C(O)=O
C10H19NO4S
MFCD00190811
boc-dl-met-oh, boc-dl-methionine, methionine,n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl methionine, 2-tert-butoxy carbonyl amino-4-methylsulfanyl butanoic acid, 2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoic acid, n-alpha-t-boc-l-methionine, n-tert-butoxycarbonyl-d-methionine, tert-butoxycarbonyl-l-methionine, pubchem11999
2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid

Specifications

93000-03-4
MFCD00190811
IMUSLIHRIYOHEV-UHFFFAOYNA-N
2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
249.33
249.33
Solid
C10H19NO4S
boc-dl-met-oh, boc-dl-methionine, methionine,n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl methionine, 2-tert-butoxy carbonyl amino-4-methylsulfanyl butanoic acid, 2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoic acid, n-alpha-t-boc-l-methionine, n-tert-butoxycarbonyl-d-methionine, tert-butoxycarbonyl-l-methionine, pubchem11999
CSCCC(NC(=O)OC(C)(C)C)C(O)=O
1 g
102827
98%
N-Boc-DL-methionine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only