missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-tert-leucine, 95%, Thermo Scientific Chemicals

Catalog No. AAH62473MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62473MD 250 mg
AAH6247303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62473MD Supplier Thermo Scientific Chemicals Supplier No. H62473MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124655-17-0
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00065575
InChI Key LRFZIPCTFBPFLX-JLDDOWRYNA-N
Synonym boc-d-tert-leucine, n-boc-d-tert-leucine, boc-d-tle-oh, r-n-tert-butoxycarbonyl-tert-leucine, r-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-d-alpha-t-butylglycine, boc-tbu-d-gly-oh, boc-d-t-leu, d-valine, n-1,1-dimethylethoxy carbonyl-3-methyl, n-tert-butoxycarbonyl-d-tert-leucine
PubChem CID 7005057
SMILES CC(C)(C)OC(=O)N[C@@H](C(O)=O)C(C)(C)C

Specifications

IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Quantity 250 mg
Formula Weight 231.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-D-tert-leucine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only