missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-serine methyl ester, 97%

Catalog No. AAH6129606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6129606 5 g
AAH6129614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6129606 Supplier Thermo Scientific Chemicals Supplier No. H6129606

CAS: 95715-85-8 | C9H17NO5 | 219.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95715-85-8
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00270516
InChI Key SANNKFASHWONFD-ZCFIWIBFSA-N
Synonym boc-d-ser-ome, boc-d-serine methyl ester, n-boc-d-serine methyl ester, n-boc-d-serine methylester, n-tert-butoxycarbonyl-d-serine methyl ester, boc-d-serinemethyl ester, n-boc-d-serinemethylester, --methyl n-boc-d-serinate, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl n-tert-butoxycarbonyl-d-serinate
PubChem CID 377723
IUPAC Name methyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

Boiling Point 215°C
Color Colorless to Yellow
Refractive Index 1.453
Density 1.080 g/mL
Quantity 5 g
Flash Point 113°C (235°F)
Formula Weight 219.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Boc-D-serine methyl ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only