missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-proline, 98+%

Catalog No. AAL0882606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0882606 5 g
AAL0882603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0882606 Supplier Thermo Scientific Chemicals Supplier No. L0882606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37784-17-1 | C10H17NO4 | 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37784-17-1
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00063226
InChI Key ZQEBQGAAWMOMAI-SSDOTTSWSA-N
Synonym n-boc-d-proline, boc-d-proline, boc-d-pro-oh, r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl-d-proline, 2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, n-t-boc-d-proline, boc-d-pro, n-alpha-t-boc-d-proline, 1-tert-butoxycarbonyl-d-proline
PubChem CID 688022
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O

Specifications

Melting Point 133°C to 135°C
Quantity 5 g
Beilstein 479316
Optical Rotation +61° (c=1 in Acetic Acid)
Formula Weight 215.25
Percent Purity ≥98%
Chemical Name or Material N-Boc-D-proline
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only