missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-pipecolinic acid, 98%

Catalog No. p-7034290
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5196106 5 g
AAH5196114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5196106 Supplier Thermo Scientific Chemicals Supplier No. H5196106
Only null left

CAS: 28697-17-8 | C11H19NO4 | 229.276 g/mol

It is also applied as catalytic agent and petrochemical additive. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also applied as catalytic agent and petrochemical additive. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Soluble in 2ml dimethyl formamide 0.3gram.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 28697-17-8
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00237380
InChI Key JQAOHGMPAAWWQO-MRVPVSSYSA-N
Synonym r-+-n-boc-2-piperidinecarboxylic acid, boc-d-hopro-oh, boc-d-pipecolic acid, r-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-d-pip-oh, d-n-boc-pipecolic acid, r-1-n-boc-pipecolinic acid, r-n-boc-piperidine-2-carboxylic acid, +-n-boc-r-pipecolinic acid, r-n-boc-2-piperidinecarboxylic acid
PubChem CID 688618
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

Melting Point 116°C to 119°C
Quantity 5 g
Beilstein 480023
Solubility Information Soluble in 2ml dimethyl formamide 0.3gram.
Optical Rotation +68°, (c=1 in Acetic Acid)
Formula Weight 229.28
Percent Purity 98%
Chemical Name or Material N-Boc-D-pipecolinic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only