Learn More
N-Boc-D-phenylglycine, 99%
CAS: 33125-05-2 | C13H17NO4 | 251.28 g/mol
$86.70 - $264.63
Chemical Identifiers
| CAS | 33125-05-2 |
|---|---|
| Molecular Formula | C13H17NO4 |
| Molecular Weight (g/mol) | 251.28 |
| MDL Number | MFCD00062043,MFCD00065588 |
| InChI Key | HOBFSNNENNQQIU-UHFFFAOYNA-N |
| Synonym | boc-d-phg-oh, boc-d-phenylglycine, n-boc-d-phenylglycine, n-boc-d-2-phenylglycine, n-boc-l-phenylglycine, n-tert-butoxycarbonyl-d-2-phenylglycine, boc-d-alpha-phenylglycine, r-tert-butoxycarbonyl amino phenyl acetic acid, r-tert-butoxycarbonylamino-phenyl-acetic acid, r-2-tert-butoxycarbonyl amino-2-phenylacetic acid |
| PubChem CID | 2755953 |
| IUPAC Name | (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid |
| SMILES | CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1854003
|
Thermo Scientific Chemicals
L1854003 |
1 g |
Each for $86.70
|
|
|||||
|
AAL1854006
|
Thermo Scientific Chemicals
L1854006 |
5 g |
Each for $264.63
|
|
|||||
Description
It is a reagent of choice for assignment of absolute configuration of chiral primary amines by 1H NMR, giving better results than Mosher's acid ((R)-(+)-Methoxy-(trifluoromethyl)ÂphenylÂacetic acid. αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is D-(+)-2-Phenylglycine with Boc protecting group. (αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is used as part of a catalyst combination to catalyze regioselective [4 + 2] cycloadditions of β-substituted cyclic enones and polyconjugated malononitriles. It can also be used to catalyze stereoselective preparation of polyfunctional nitrocyclohexene carboxaldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a reagent of choice for assignment of absolute configuration of chiral primary amines by 1H NMR, giving better results than Mosher′s acid ((R)-(+)-É-Methoxy-É-(trifluoromethyl)ÂphenylÂacetic acid. αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is D-(+)-2-Phenylglycine with Boc protecting group. (αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is used as part of a catalyst combination to catalyze regioselective [4 + 2] cycloadditions of β-substituted cyclic enones and polyconjugated malononitriles. It can also be used to catalyze stereoselective preparation of polyfunctional nitrocyclohexene carboxaldehydes.
Solubility
Insoluble in water. Slightly soluble in DMSO and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 33125-05-2 | |
| 251.28 | |
| HOBFSNNENNQQIU-UHFFFAOYNA-N | |
| 2755953 | |
| CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1 |
| C13H17NO4 | |
| MFCD00062043,MFCD00065588 | |
| boc-d-phg-oh, boc-d-phenylglycine, n-boc-d-phenylglycine, n-boc-d-2-phenylglycine, n-boc-l-phenylglycine, n-tert-butoxycarbonyl-d-2-phenylglycine, boc-d-alpha-phenylglycine, r-tert-butoxycarbonyl amino phenyl acetic acid, r-tert-butoxycarbonylamino-phenyl-acetic acid, r-2-tert-butoxycarbonyl amino-2-phenylacetic acid | |
| (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid |
Specifications
| 90°C to 92°C | |
| C13H17NO4 | |
| 3033982 | |
| Insoluble in water. Slightly soluble in DMSO and methanol. | |
| CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1 | |
| 1 g | |
| 2755953 | |
| 251.28 | |
| N-Boc-D-phenylglycine |
| 33125-05-2 | |
| MFCD00062043,MFCD00065588 | |
| boc-d-phg-oh, boc-d-phenylglycine, n-boc-d-phenylglycine, n-boc-d-2-phenylglycine, n-boc-l-phenylglycine, n-tert-butoxycarbonyl-d-2-phenylglycine, boc-d-alpha-phenylglycine, r-tert-butoxycarbonyl amino phenyl acetic acid, r-tert-butoxycarbonylamino-phenyl-acetic acid, r-2-tert-butoxycarbonyl amino-2-phenylacetic acid | |
| HOBFSNNENNQQIU-UHFFFAOYNA-N | |
| (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid | |
| 251.28 | |
| −144° (c=1 in Ethanol) | |
| 99% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only