missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog No. AAH2771303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2771303 1 g
AAH2771306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2771303 Supplier Thermo Scientific Chemicals Supplier No. H2771303

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106454-69-7
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.33
MDL Number MFCD00216472
InChI Key LDKDMDVMMCXTMO-GFCCVEGCSA-N
Synonym n-boc-d-phenylalaninol, boc-d-phenylalaninol, r-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, n-tert-butoxycarbonyl-d-phenylalaninol, r-2-boc-amino-3-phenyl-1-propanol, r-+-2-tert-butoxycarbonylamino-3-phenyl-1-propanol, r-2-tert-butoxycarbonylamino-3-phenyl-1-propanol, tert-butyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate, pubchem5739, r-n-tert-butoxycarbonyl-beta-phenylalaninol
PubChem CID 7019371
IUPAC Name tert-butyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 94°C to 96°C
Quantity 1 g
Formula Weight 251.33
Percent Purity 98%
Chemical Name or Material N-Boc-D-phenylalaninol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only