missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-D-Phenylalaninal, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 77119-85-8 |
|---|---|
| Molecular Formula | C14H19NO3 |
| Molecular Weight (g/mol) | 249.31 |
| MDL Number | MFCD00274187,MFCD00801183 |
| InChI Key | ZJTYRNPLVNMVPQ-GFCCVEGCSA-N |
| Synonym | n-boc-d-phenylalaninal, boc-d-phenylalaninal, r-+-2-tert-butoxycarbonylamino-3-phenylpropanal, r-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, n-boc-2 r-3-phenylpropanal, tert-butyl n-2r-1-oxo-3-phenylpropan-2-yl carbamate, tert-butyl 2r-1-oxo-3-phenylpropan-2-yl carbamate, carbamic acid, 1r-1-formyl-2-phenylethyl-, 1,1-dimethylethyl ester, boc-d-phe-h, pubchem11217 |
| PubChem CID | 7020444 |
| IUPAC Name | tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate |
| SMILES | CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC376732500
|
N/A | 250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 77119-85-8 | |
| 249.31 | |
| ZJTYRNPLVNMVPQ-GFCCVEGCSA-N | |
| 7020444 | |
| CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O |
| C14H19NO3 | |
| MFCD00274187,MFCD00801183 | |
| n-boc-d-phenylalaninal, boc-d-phenylalaninal, r-+-2-tert-butoxycarbonylamino-3-phenylpropanal, r-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, n-boc-2 r-3-phenylpropanal, tert-butyl n-2r-1-oxo-3-phenylpropan-2-yl carbamate, tert-butyl 2r-1-oxo-3-phenylpropan-2-yl carbamate, carbamic acid, 1r-1-formyl-2-phenylethyl-, 1,1-dimethylethyl ester, boc-d-phe-h, pubchem11217 | |
| tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate |
Specifications
| 77119-85-8 | |
| 100.0 | |
| 96% min. (GC) | |
| MFCD00274187,MFCD00801183 | |
| n-boc-d-phenylalaninal, boc-d-phenylalaninal, r-+-2-tert-butoxycarbonylamino-3-phenylpropanal, r-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, n-boc-2 r-3-phenylpropanal, tert-butyl n-2r-1-oxo-3-phenylpropan-2-yl carbamate, tert-butyl 2r-1-oxo-3-phenylpropan-2-yl carbamate, carbamic acid, 1r-1-formyl-2-phenylethyl-, 1,1-dimethylethyl ester, boc-d-phe-h, pubchem11217 | |
| CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O | |
| 249.31 | |
| 249.31 | |
| N-BOC-D-Phenylalaninal |
| 96.0 | |
| Authentic | |
| C14H19NO3 | |
| + 45.00 | |
| ZJTYRNPLVNMVPQ-GFCCVEGCSA-N | |
| tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate | |
| 7020444 | |
| 97% |
RUO – Research Use Only