missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-leucine hydrate, 98+%

Catalog No. AAL0912403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0912403 1 g
AAL0912406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0912403 Supplier Thermo Scientific Chemicals Supplier No. L0912403

CAS: 16937-99-8 | C11H23NO5 | 249.307 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16937-99-8
Molecular Formula C11H23NO5
Molecular Weight (g/mol) 249.307
MDL Number MFCD00065583
InChI Key URQQEIOTRWJXBA-DDWIOCJRSA-N
Synonym boc-d-leucine monohydrate, r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leu-oh.h2o, n-boc-d-leucine monohydrate, boc-d-leu-oh h2o, d-leucine, n-1,1-dimethylethoxy carbonyl-, monohydrate, 2r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leucine h2o, boc-d-leucine hydrate, n-boc-d-leucinemonohydrate
PubChem CID 2724763
IUPAC Name (2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate
SMILES CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C.O

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
Beilstein 2331060
Optical Rotation +25° (c=2 in Acetic Acid)
Formula Weight 231.29 (Anhydrous)
Percent Purity ≥98%
Chemical Name or Material N-Boc-D-leucine hydrate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only