missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
$146.25
Chemical Identifiers
| CAS | 104719-63-3 |
|---|---|
| Molecular Formula | C14H25NO6 |
| Molecular Weight (g/mol) | 303.355 |
| MDL Number | MFCD00076927 |
| InChI Key | YGSRAYJBEREVRB-SECBINFHSA-N |
| Synonym | boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester |
| PubChem CID | 7018748 |
| IUPAC Name | (2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
| SMILES | CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH6317206
|
N/A | 5 g |
Each for $146.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 104719-63-3 | |
| 303.355 | |
| YGSRAYJBEREVRB-SECBINFHSA-N | |
| 7018748 | |
| CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C |
| C14H25NO6 | |
| MFCD00076927 | |
| boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester | |
| (2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
Specifications
| 104719-63-3 | |
| MFCD00076927 | |
| boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester | |
| CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C | |
| 303.355 | |
| 303.35 | |
| Solid |
| C14H25NO6 | |
| 3594191 | |
| YGSRAYJBEREVRB-SECBINFHSA-N | |
| (2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid | |
| 7018748 | |
| 98% | |
| N-Boc-D-glutamic acid 5-tert-butyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only