missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog No. aah6317206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6317206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6317206 Supplier Thermo Scientific Chemicals Supplier No. H6317206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104719-63-3 | Molecular Formula: C14H25NO6 | Molecular Weight: 303.355 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104719-63-3
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.355
MDL Number MFCD00076927
InChI Key YGSRAYJBEREVRB-SECBINFHSA-N
Synonym boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester
PubChem CID 7018748
IUPAC Name (2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Beilstein 3594191
Quantity 5 g
Formula Weight 303.35
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-D-glutamic acid 5-tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only